Search
Search

CCAR2| Gene Symbol: CCAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194900.3 | NP_001181829.1 | 606 | 65 | ||
| XM_001106923.4 | XP_001106923.2 | 399 | 65 | |||
| XM_015144902.2 | XP_015000388.1 | 399 | 65 | |||
| GenBank mRNA | JU335685.1 | - | 517 | 65 | ||
| JU335686.1 | - | 517 | 65 | |||
| JU476586.1 | - | 192 | 65 | |||
| JU476587.1 | - | 192 | 65 | |||
| JV047784.1 | - | 491 | 65 | |||
| JV047785.1 | - | 491 | 65 | |||
| JV624228.1 | - | 522 | 65 | |||
| JV657491.1 | - | 583 | 65 | |||
Set Membership: |
|