Search
Search

EIF2B4| Gene Symbol: EIF2B4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111967.2 | XP_014967453.2 | 1232 | 89 | ||
| XM_015111968.2 | XP_014967454.2 | 1204 | 89 | |||
| XM_015111969.2 | XP_014967455.2 | 1249 | 89 | |||
| XM_015111970.2 | XP_014967456.2 | 1234 | 89 | |||
| XM_028831841.1 | XP_028687674.1 | 1233 | 89 | |||
| XM_028831842.1 | XP_028687675.1 | 1231 | 89 | |||
| XM_028831843.1 | XP_028687676.1 | 1202 | 89 | |||
| XM_028831844.1 | XP_028687677.1 | 1230 | 89 | |||
| XM_028831845.1 | XP_028687678.1 | 1178 | 89 | |||
| XM_028831846.1 | XP_028687679.1 | 1176 | 89 | |||
| XM_028831847.1 | XP_028687680.1 | 1025 | 89 | |||
| XM_028831848.1 | XP_028687681.1 | 945 | 89 | |||
| GenBank mRNA | JU321258.1 | - | 1149 | 89 | ||
| JU472101.1 | - | 1092 | 89 | |||
| JV043844.1 | - | 1033 | 89 | |||
| JV527775.1 | - | 1028 | 89 | |||
| JV602338.1 | - | 1117 | 89 | |||
Set Membership: |
|