Search
Search

KCNH2| Gene Symbol: KCNH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135279.2 | XP_014990765.2 | 3731 | 60 | ||
| XM_015135282.2 | XP_014990768.1 | 2705 | 60 | |||
| GenBank mRNA | JU328043.1 | - | 573 | 60 | ||
| JV043362.1 | - | 3347 | 60 | |||
| JV043363.1 | - | 2587 | 60 | |||
| JV636139.1 | - | 3351 | 60 | |||
| JV636140.1 | - | 2615 | 60 | |||
| JV714379.1 | - | 596 | 60 | |||
Set Membership: |
|