Search
Search

PTPRA| Gene Symbol: PTPRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114682.4 | XP_001114682.2 | 2477 | 89 | ||
| XM_002798218.3 | XP_002798264.2 | 2461 | 89 | |||
| XM_015149415.2 | XP_015004901.2 | 2504 | 89 | |||
| XM_015149418.2 | XP_015004904.2 | 2386 | 89 | |||
| XM_028828269.1 | XP_028684102.1 | 2082 | 89 | |||
| XM_028828270.1 | XP_028684103.1 | 2055 | 89 | |||
| XM_028828271.1 | XP_028684104.1 | 2099 | 89 | |||
| XM_028828272.1 | XP_028684105.1 | 1325 | 89 | |||
| GenBank mRNA | AF424826.1 | - | 306 | 89 | ||
| JU335690.1 | - | 2298 | 89 | |||
| JU335691.1 | - | 2271 | 89 | |||
| JU476589.1 | - | 2279 | 89 | |||
| JU476590.1 | - | 2252 | 89 | |||
| JV047790.1 | - | 2218 | 89 | |||
| JV657520.1 | - | 2407 | 89 | |||
Set Membership: |
|