Search
Search

CAPN2| Gene Symbol: CAPN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260667.1 | NP_001247596.1 | 2213 | 103 | ||
| GenBank mRNA | JU320808.1 | - | 2271 | 103 | ||
| JU477389.1 | - | 2100 | 103 | |||
| JV044828.1 | - | 2213 | 103 | |||
| JV630183.1 | - | 976 | 103 | |||
Set Membership: |
|