Search
Search

TNFRSF19| Gene Symbol: TNFRSF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002808139.3 | XP_002808185.2 | 813 | 58 | ||
| XM_015120742.2 | XP_014976228.1 | 401 | 58 | |||
| XM_015120743.2 | XP_014976229.1 | 813 | 58 | |||
| XM_015120744.2 | XP_014976230.1 | 401 | 58 | |||
| XM_015120745.2 | XP_014976231.1 | 813 | 58 | |||
| XM_028837411.1 | XP_028693244.1 | 359 | 58 | |||
| XM_028837412.1 | XP_028693245.1 | 374 | 58 | |||
| GenBank mRNA | JU324776.1 | - | 228 | 58 | ||
| JV065177.1 | - | 281 | 58 | |||
| JV473723.1 | - | 269 | 58 | |||
Set Membership: |
|