Search
Search

NUP37| Gene Symbol: NUP37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260738.1 | NP_001247667.1 | 926 | 75 | ||
| XM_015152529.2 | XP_015008015.1 | 1124 | 75 | |||
| XM_015152531.2 | XP_015008017.1 | 734 | 75 | |||
| GenBank mRNA | JU320879.1 | - | 926 | 75 | ||
| JU472939.1 | - | 921 | 75 | |||
Set Membership: |
|