Search
Search

PLBD2| Gene Symbol: PLBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152887.2 | XP_015008373.2 | 1318 | 61 | ||
| GenBank mRNA | JU325644.1 | - | 1244 | 61 | ||
| JU470610.1 | - | 1294 | 61 | |||
| JV573693.1 | - | 1319 | 61 | |||
| JV667358.1 | - | 1320 | 61 | |||
Set Membership: |
|