Search
Search

ARHGEF3| Gene Symbol: ARHGEF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101044.4 | XP_001101044.3 | 989 | 82 | ||
| XM_001101218.4 | XP_001101218.3 | 2723 | 82 | |||
| XM_001101312.4 | XP_001101312.2 | 1196 | 82 | |||
| XM_015130642.2 | XP_014986128.1 | 1071 | 82 | |||
| XM_028844283.1 | XP_028700116.1 | 1329 | 82 | |||
| XM_028844284.1 | XP_028700117.1 | 1237 | 82 | |||
| XM_028844285.1 | XP_028700118.1 | 916 | 82 | |||
| XM_028844286.1 | XP_028700119.1 | 2607 | 82 | |||
| GenBank mRNA | JU327320.1 | - | 893 | 82 | ||
| JU334734.1 | - | 996 | 82 | |||
| JU475785.1 | - | 903 | 82 | |||
| JV045256.1 | - | 944 | 82 | |||
| JV566559.1 | - | 1006 | 82 | |||
| JV636187.1 | - | 1013 | 82 | |||
Set Membership: |
|