Search
Search

SLC35D1| Gene Symbol: SLC35D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258070.1 | NP_001244999.1 | 780 | 86 | ||
| XM_015141577.2 | XP_014997063.1 | 775 | 86 | |||
| XM_015141585.2 | XP_014997071.1 | 777 | 86 | |||
| GenBank mRNA | JU471272.1 | - | 780 | 86 | ||
Set Membership: |
|