Search
Search

CASP8AP2| Gene Symbol: CASP8AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847347.1 | XP_028703180.1 | 6096 | 123 | ||
| XM_028847348.1 | XP_028703181.1 | 6172 | 123 | |||
| XM_028847349.1 | XP_028703182.1 | 6128 | 123 | |||
| XM_028847350.1 | XP_028703183.1 | 6122 | 123 | |||
| XM_028847351.1 | XP_028703184.1 | 6361 | 123 | |||
| XM_028847352.1 | XP_028703185.1 | 6021 | 123 | |||
| XM_028847353.1 | XP_028703186.1 | 6173 | 123 | |||
| XM_028847354.1 | XP_028703187.1 | 6189 | 123 | |||
| XM_028847355.1 | XP_028703188.1 | 6056 | 123 | |||
| XM_028847357.1 | XP_028703190.1 | 6295 | 123 | |||
| XM_028847358.1 | XP_028703191.1 | 6031 | 123 | |||
| GenBank mRNA | JU337405.1 | - | 6140 | 123 | ||
| JU478118.1 | - | 6154 | 123 | |||
Set Membership: |
|