Search
Search

DHX33| Gene Symbol: DHX33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118388.2 | XP_014973874.1 | 1339 | 64 | ||
| XM_015118389.2 | XP_014973875.1 | 1179 | 64 | |||
| GenBank mRNA | JU326964.1 | - | 996 | 64 | ||
| JV719115.1 | - | 1045 | 64 | |||
| JV719116.1 | - | 1045 | 64 | |||
Set Membership: |
|