Search
Search

SLC12A2| Gene Symbol: SLC12A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261785.1 | NP_001248714.1 | 2776 | 71 | ||
| XM_015140677.2 | XP_014996163.1 | 2766 | 71 | |||
| GenBank mRNA | JU336002.1 | - | 2776 | 71 | ||
| JU336003.1 | - | 2776 | 71 | |||
| JU336004.1 | - | 2776 | 71 | |||
| JU336005.1 | - | 2776 | 71 | |||
| JU336006.1 | - | 2776 | 71 | |||
| JU336007.1 | - | 2776 | 71 | |||
Set Membership: |
|