Search
Search

ZNF641| Gene Symbol: ZNF641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001096706.4 | XP_001096706.3 | 335 | 89 | ||
| XM_015151568.2 | XP_015007054.2 | 720 | 89 | |||
| XM_028829436.1 | XP_028685269.1 | 929 | 89 | |||
| XM_028829437.1 | XP_028685270.1 | 311 | 89 | |||
Set Membership: |
|