Search
Search

INF2| Gene Symbol: INF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144594.2 | XP_015000080.2 | 1825 | 55 | ||
| XM_028851934.1 | XP_028707767.1 | 1883 | 55 | |||
| GenBank mRNA | JU347174.1 | - | 352 | 55 | ||
Set Membership: |
|