Search
Search

CDC16| Gene Symbol: CDC16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266466.1 | NP_001253395.1 | 327 | 61 | ||
| XM_015121537.2 | XP_014977023.1 | 450 | 61 | |||
| XM_015121538.2 | XP_014977024.1 | 450 | 61 | |||
| XM_015121540.2 | XP_014977026.1 | 96 | 61 | |||
| GenBank mRNA | JU336936.1 | - | 298 | 61 | ||
| JU336937.1 | - | 298 | 61 | |||
| JU477670.1 | - | 327 | 61 | |||
| JU477671.1 | - | 327 | 61 | |||
| JV632396.1 | - | 340 | 61 | |||
Set Membership: |
|