Search
Search

GNPAT| Gene Symbol: GNPAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265780.1 | NP_001252709.1 | 1843 | 139 | ||
| XM_028844822.1 | XP_028700655.1 | 1883 | 139 | |||
| XM_028844825.1 | XP_028700658.1 | 1881 | 139 | |||
| GenBank mRNA | JU322776.1 | - | 1901 | 139 | ||
| JU472255.1 | - | 1823 | 139 | |||
| JV045491.1 | - | 1902 | 139 | |||
| JV565033.1 | - | 1843 | 139 | |||
| JV636996.1 | - | 1905 | 139 | |||
Set Membership: |
|