Search
Search

KDM2B| Gene Symbol: KDM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152991.2 | XP_015008477.2 | 3883 | 89 | ||
| XM_015152992.2 | XP_015008478.2 | 3874 | 89 | |||
| XM_015152993.2 | XP_015008479.2 | 3894 | 89 | |||
| XM_015152994.2 | XP_015008480.2 | 3884 | 89 | |||
| XM_015152995.2 | XP_015008481.2 | 3395 | 89 | |||
| XM_015152996.2 | XP_015008482.2 | 3769 | 89 | |||
| XM_015152997.2 | XP_015008483.2 | 3386 | 89 | |||
| XM_015152998.2 | XP_015008484.2 | 3771 | 89 | |||
| XM_015152999.2 | XP_015008485.2 | 3591 | 89 | |||
| XM_015153000.2 | XP_015008486.2 | 3426 | 89 | |||
| XM_015153001.2 | XP_015008487.2 | 3112 | 89 | |||
| XM_015153002.2 | XP_015008488.2 | 3575 | 89 | |||
| XM_015153003.2 | XP_015008489.2 | 3262 | 89 | |||
| XM_028830002.1 | XP_028685835.1 | 3760 | 89 | |||
| XM_028830003.1 | XP_028685836.1 | 3121 | 89 | |||
| XM_028830004.1 | XP_028685837.1 | 3009 | 89 | |||
| GenBank mRNA | JU327218.1 | - | 1514 | 89 | ||
Set Membership: |
|