Search
Search

BCO2| Gene Symbol: BCO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115695.2 | XP_014971181.1 | 1234 | 112 | ||
| XM_015115696.2 | XP_014971182.1 | 1120 | 112 | |||
| XM_028833968.1 | XP_028689801.1 | 1234 | 112 | |||
| XM_028833969.1 | XP_028689802.1 | 1234 | 112 | |||
| GenBank mRNA | JU336637.1 | - | 1431 | 112 | ||
Set Membership: |
|