Search
Search

GPR15| Gene Symbol: GPR15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042647.1 | NP_001036112.1 | 831 | 60 | ||
| GenBank mRNA | AF084226.1 | - | 831 | 60 | ||
| AF084227.1 | - | 831 | 60 | |||
| DQ887987.1 | - | 831 | 60 | |||
| DQ887990.1 | - | 305 | 60 | |||
| DQ887992.1 | - | 831 | 60 | |||
| DQ887993.1 | - | 831 | 60 | |||
| DQ887994.1 | - | 831 | 60 | |||
| DQ887996.1 | - | 831 | 60 | |||
| DQ887998.1 | - | 831 | 60 | |||
| DQ888003.1 | - | 831 | 60 | |||
| DQ888005.1 | - | 784 | 60 | |||
| DQ888006.1 | - | 831 | 60 | |||
| DQ888008.1 | - | 277 | 60 | |||
| DQ888009.1 | - | 786 | 60 | |||
| DQ888011.1 | - | 304 | 60 | |||
| DQ888012.1 | - | 831 | 60 | |||
| DQ888013.1 | - | 831 | 60 | |||
| DQ888014.1 | - | 276 | 60 | |||
| DQ888016.1 | - | 831 | 60 | |||
| DQ888017.1 | - | 831 | 60 | |||
| DQ888019.1 | - | 303 | 60 | |||
| DQ888021.1 | - | 275 | 60 | |||
| DQ888022.1 | - | 784 | 60 | |||
| DQ888023.1 | - | 777 | 60 | |||
| DQ888024.1 | - | 831 | 60 | |||
| DQ888026.1 | - | 831 | 60 | |||
| DQ888027.1 | - | 831 | 60 | |||
| DQ888028.1 | - | 831 | 60 | |||
| DQ888029.1 | - | 831 | 60 | |||
| DQ888030.1 | - | 786 | 60 | |||
| DQ888033.1 | - | 276 | 60 | |||
| DQ888036.1 | - | 831 | 60 | |||
| DQ888037.1 | - | 831 | 60 | |||
| DQ888038.1 | - | 831 | 60 | |||
Set Membership: |
|