Search
Search

TSACC| Gene Symbol: TSACC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112034.2 | XP_014967520.1 | 321 | 84 | ||
| XM_015112041.2 | XP_014967527.1 | 220 | 84 | |||
| XM_015112048.2 | XP_014967534.1 | 328 | 84 | |||
| XM_015112054.2 | XP_014967540.1 | 227 | 84 | |||
Set Membership: |
|