Search
Search

SUCLA2| Gene Symbol: SUCLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121053.2 | XP_014976539.2 | 879 | 151 | ||
| GenBank mRNA | JU322973.1 | - | 835 | 151 | ||
| JU472546.1 | - | 825 | 151 | |||
| JV043945.1 | - | 826 | 151 | |||
| JV602325.1 | - | 852 | 151 | |||
Set Membership: |
|