Search
Search

PSMA6| Gene Symbol: PSMA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193553.1 | NP_001180482.1 | 79 | 119 | ||
| XM_028850201.1 | XP_028706034.1 | 171 | 119 | |||
| GenBank mRNA | CD767409.1 | - | 83 | 119 | ||
| JU320666.1 | - | 103 | 119 | |||
| JU472443.1 | - | 123 | 119 | |||
| JV045267.1 | - | 110 | 119 | |||
Set Membership: |
|