Search
Search

CTHRC1| Gene Symbol: CTHRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085750.4 | XP_001085750.2 | 763 | 142 | ||
| XM_015145808.2 | XP_015001294.2 | 771 | 142 | |||
| GenBank mRNA | JU331146.1 | - | 516 | 142 | ||
| JV570541.1 | - | 609 | 142 | |||
Set Membership: |
|