Search
Search

SGSH| Gene Symbol: SGSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001110101.4 | XP_001110101.4 | 172 | 87 | ||
| XM_015120506.2 | XP_014975992.2 | 172 | 87 | |||
| XM_028836882.1 | XP_028692715.1 | 172 | 87 | |||
| GenBank mRNA | JV637406.1 | - | 123 | 87 | ||
Set Membership: |
|