Search
Search

LAT| Gene Symbol: LAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125849.2 | XP_014981335.1 | 905 | 98 | ||
| XM_015125850.2 | XP_014981336.1 | 744 | 98 | |||
| XM_028841025.1 | XP_028696858.1 | 825 | 98 | |||
Set Membership: |
|