Search
Search

DCLK2| Gene Symbol: DCLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082020.4 | XP_001082020.3 | 1414 | 72 | ||
| XM_015139326.2 | XP_014994812.2 | 2339 | 72 | |||
| XM_015139327.2 | XP_014994813.2 | 1465 | 72 | |||
| XM_015139328.2 | XP_014994814.2 | 2168 | 72 | |||
| XM_015139329.2 | XP_014994815.2 | 1468 | 72 | |||
| XM_028848815.1 | XP_028704648.1 | 1468 | 72 | |||
| GenBank mRNA | JU321421.1 | - | 1519 | 72 | ||
Set Membership: |
|