Search
Search

CNDP1| Gene Symbol: CNDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085457.4 | XP_001085457.2 | 1549 | 72 | ||
| XM_015122203.2 | XP_014977689.1 | 1377 | 72 | |||
| GenBank mRNA | JU336771.1 | - | 1512 | 72 | ||
| JV725380.1 | - | 1512 | 72 | |||
Set Membership: |
|