Search
Search

CHDH| Gene Symbol: CHDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257809.1 | NP_001244738.1 | 134 | 103 | ||
| XM_015130537.2 | XP_014986023.1 | 96 | 103 | |||
| XM_028843080.1 | XP_028698913.1 | 350 | 103 | |||
| GenBank mRNA | JU335324.1 | - | 134 | 103 | ||
| JU335325.1 | - | 134 | 103 | |||
| JU335326.1 | - | 134 | 103 | |||
| JU335327.1 | - | 134 | 103 | |||
| JV688988.1 | - | 173 | 103 | |||
Set Membership: |
|