Search
Search

UPRT| Gene Symbol: UPRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261749.1 | NP_001248678.1 | 516 | 80 | ||
| XM_015127698.2 | XP_014983184.1 | 614 | 80 | |||
| XM_028841768.1 | XP_028697601.1 | 618 | 80 | |||
| GenBank mRNA | JU474360.1 | - | 516 | 80 | ||
Set Membership: |
|