Search
Search

PEX13| Gene Symbol: PEX13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258170.1 | NP_001245099.1 | 871 | 98 | ||
| GenBank mRNA | JU321132.1 | - | 814 | 98 | ||
| JU473799.1 | - | 871 | 98 | |||
Set Membership: |
|