Search
Search

CSTF2T| Gene Symbol: CSTF2T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266798.1 | NP_001253727.1 | 1325 | 106 | ||
| GenBank mRNA | JU474885.1 | - | 1324 | 106 | ||
| JV046553.1 | - | 1317 | 106 | |||
| JV636056.1 | - | 1325 | 106 | |||
Set Membership: |
|