Search
Search

HERPUD2| Gene Symbol: HERPUD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133722.2 | XP_014989208.1 | 1133 | 76 | ||
| XM_028845866.1 | XP_028701699.1 | 364 | 76 | |||
| GenBank mRNA | JU335887.1 | - | 921 | 76 | ||
| JU335888.1 | - | 921 | 76 | |||
| JU335889.1 | - | 921 | 76 | |||
| JU335890.1 | - | 921 | 76 | |||
| JU335891.1 | - | 921 | 76 | |||
| JU335892.1 | - | 921 | 76 | |||
| JU335893.1 | - | 921 | 76 | |||
| JU335894.1 | - | 921 | 76 | |||
| JU335895.1 | - | 921 | 76 | |||
| JU335896.1 | - | 921 | 76 | |||
| JU471656.1 | - | 1143 | 76 | |||
| JU471657.1 | - | 1143 | 76 | |||
| JV043995.1 | - | 1146 | 76 | |||
| JV638781.1 | - | 1147 | 76 | |||