Search
Search

PM20D1| Gene Symbol: PM20D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119095.2 | XP_014974581.2 | 1021 | 97 | ||
| XM_028852495.1 | XP_028708328.1 | 1040 | 97 | |||
| GenBank mRNA | JV643247.1 | - | 149 | 97 | ||
Set Membership: |
|