Search
Search

GYPC| Gene Symbol: GYPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015109882.2 | XP_014965368.1 | 551 | 60 | ||
| XM_028830629.1 | XP_028686462.1 | 656 | 60 | |||
| XM_028830631.1 | XP_028686464.1 | 557 | 60 | |||
| GenBank mRNA | JV059239.1 | - | 44 | 60 | ||
| JV649581.1 | - | 38 | 60 | |||
Set Membership: |
|