Search
Search

MVK| Gene Symbol: MVK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152731.2 | XP_015008217.1 | 903 | 89 | ||
| XM_028829844.1 | XP_028685677.1 | 903 | 89 | |||
| GenBank mRNA | JU323111.1 | - | 791 | 89 | ||
| JU323112.1 | - | 791 | 89 | |||
| JU472020.1 | - | 793 | 89 | |||
| JV045781.1 | - | 794 | 89 | |||
| JV639472.1 | - | 810 | 89 | |||
Set Membership: |
|