Search
Search

ITCH| Gene Symbol: ITCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149167.1 | XP_015004653.1 | 2303 | 94 | ||
| XM_015149170.2 | XP_015004656.1 | 2146 | 94 | |||
| XM_015149171.2 | XP_015004657.1 | 2055 | 94 | |||
| XM_015149172.2 | XP_015004658.1 | 2225 | 94 | |||
| XM_015149173.1 | XP_015004659.1 | 1028 | 94 | |||
| XM_028828339.1 | XP_028684172.1 | 2447 | 94 | |||
| GenBank mRNA | JU321209.1 | - | 2274 | 94 | ||
| JU332142.1 | - | 2274 | 94 | |||
| JU473364.1 | - | 2248 | 94 | |||
| JV044271.1 | - | 2213 | 94 | |||
| JV724705.1 | - | 2253 | 94 | |||
Set Membership: |
|