Search
Search

SLC35B1| Gene Symbol: SLC35B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119333.2 | XP_014974819.1 | 666 | 70 | ||
| XM_015119335.2 | XP_014974821.1 | 638 | 70 | |||
| XM_028836387.1 | XP_028692220.1 | 322 | 70 | |||
| XM_028836388.1 | XP_028692221.1 | 461 | 70 | |||
| GenBank mRNA | JU319813.1 | - | 426 | 70 | ||
| JU473041.1 | - | 405 | 70 | |||
| JV044292.1 | - | 417 | 70 | |||
| JV705028.1 | - | 401 | 70 | |||
Set Membership: |
|