Search
Search

CDC42BPB| Gene Symbol: CDC42BPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144535.2 | XP_015000021.2 | 2874 | 59 | ||
| XM_028851884.1 | XP_028707717.1 | 2876 | 59 | |||
| XM_028851885.1 | XP_028707718.1 | 2436 | 59 | |||
| XM_028851886.1 | XP_028707719.1 | 2876 | 59 | |||
| XM_028851887.1 | XP_028707720.1 | 2876 | 59 | |||
| GenBank mRNA | JU320543.1 | - | 2673 | 59 | ||
| JV048782.1 | - | 2518 | 59 | |||
| JV599099.1 | - | 2543 | 59 | |||
| JV638022.1 | - | 2712 | 59 | |||
Set Membership: |
|