Search
Search

CEP95| Gene Symbol: CEP95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001116599.4 | XP_001116599.1 | 782 | 83 | ||
| XM_015120113.2 | XP_014975599.1 | 782 | 83 | |||
| XM_028836668.1 | XP_028692501.1 | 782 | 83 | |||
| GenBank mRNA | JU325385.1 | - | 557 | 83 | ||
| JU477746.1 | - | 83 | ||||
| JV665120.1 | - | 631 | 83 | |||