Search
Search

LAMA4| Gene Symbol: LAMA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137555.2 | XP_014993041.2 | 2072 | 130 | ||
| XM_015137556.2 | XP_014993042.2 | 2047 | 130 | |||
| XM_028847051.1 | XP_028702884.1 | 1809 | 130 | |||
| GenBank mRNA | JV044400.1 | - | 1953 | 130 | ||
| JV044401.1 | - | 1932 | 130 | |||
| JV635678.1 | - | 1948 | 130 | |||
| JV635679.1 | - | 1927 | 130 | |||
Set Membership: |
|