Search
Search

MYCBP2| Gene Symbol: MYCBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121229.2 | XP_014976715.2 | 7531 | 145 | ||
| XM_015121232.2 | XP_014976718.2 | 7531 | 145 | |||
| XM_015121233.2 | XP_014976719.2 | 7423 | 145 | |||
| XM_015121234.2 | XP_014976720.2 | 7531 | 145 | |||
| XM_015121243.2 | XP_014976729.2 | 7530 | 145 | |||
| XM_015121248.2 | XP_014976734.1 | 7449 | 145 | |||
| XM_028837600.1 | XP_028693433.1 | 7531 | 145 | |||
| XM_028837601.1 | XP_028693434.1 | 7531 | 145 | |||
| XM_028837602.1 | XP_028693435.1 | 7531 | 145 | |||
| XM_028837603.1 | XP_028693436.1 | 4457 | 145 | |||
| XM_028837604.1 | XP_028693437.1 | 7530 | 145 | |||
| GenBank mRNA | JU327582.1 | - | 4570 | 145 | ||
| JU340718.1 | - | 4570 | 145 | |||
Set Membership: |
|