Search
Search

MEGF11| Gene Symbol: MEGF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142520.2 | XP_014998006.2 | 4092 | 134 | ||
| XM_015142521.2 | XP_014998007.2 | 3862 | 134 | |||
| XM_028851052.1 | XP_028706885.1 | 3963 | 134 | |||
| GenBank mRNA | JU349552.1 | - | 324 | 134 | ||
Set Membership: |
|