Search
Search

SLC12A7| Gene Symbol: SLC12A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139696.2 | XP_014995182.2 | 2504 | 98 | ||
| XM_028849316.1 | XP_028705149.1 | 2521 | 98 | |||
| XM_028849317.1 | XP_028705150.1 | 2521 | 98 | |||
| XM_028849318.1 | XP_028705151.1 | 2498 | 98 | |||
| XM_028849319.1 | XP_028705152.1 | 2348 | 98 | |||
| XM_028849320.1 | XP_028705153.1 | 2348 | 98 | |||
| XM_028849321.1 | XP_028705154.1 | 3743 | 98 | |||
| GenBank mRNA | JU479010.1 | - | 1848 | 98 | ||
| JV640541.1 | - | 2402 | 98 | |||
Set Membership: |
|