Search
Search

FADS3| Gene Symbol: FADS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002799527.3 | XP_002799573.2 | 731 | 89 | ||
| GenBank mRNA | JU326887.1 | - | 514 | 89 | ||
| JV045552.1 | - | 529 | 89 | |||
| JV638096.1 | - | 643 | 89 | |||
| JV667741.1 | - | 652 | 89 | |||