Search
Search

NUP160| Gene Symbol: NUP160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114265.2 | XP_014969751.2 | 4112 | 111 | ||
| XM_015114266.2 | XP_014969752.2 | 3684 | 111 | |||
| GenBank mRNA | JU327190.1 | - | 3679 | 111 | ||
| JU483271.1 | - | 3905 | 111 | |||
| JV646496.1 | - | 3908 | 111 | |||
Set Membership: |
|