Search
Search

KIF1A| Gene Symbol: KIF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028831325.1 | XP_028687158.1 | 1939 | 76 | ||
| XM_028831326.1 | XP_028687159.1 | 1939 | 76 | |||
| XM_028831327.1 | XP_028687160.1 | 1939 | 76 | |||
| XM_028831328.1 | XP_028687161.1 | 1939 | 76 | |||
| XM_028831329.1 | XP_028687162.1 | 1939 | 76 | |||
| XM_028831330.1 | XP_028687163.1 | 1939 | 76 | |||
| XM_028831331.1 | XP_028687164.1 | 1864 | 76 | |||
| XM_028831332.1 | XP_028687165.1 | 1837 | 76 | |||
| XM_028831333.1 | XP_028687166.1 | 1863 | 76 | |||
| XM_028831334.1 | XP_028687167.1 | 1938 | 76 | |||
| XM_028831335.1 | XP_028687168.1 | 1938 | 76 | |||
| XM_028831336.1 | XP_028687169.1 | 1938 | 76 | |||
| XM_028831337.1 | XP_028687170.1 | 1863 | 76 | |||
| XM_028831338.1 | XP_028687171.1 | 1863 | 76 | |||
| XM_028831339.1 | XP_028687172.1 | 1836 | 76 | |||
| XM_028831340.1 | XP_028687173.1 | 1836 | 76 | |||
| XM_028831341.1 | XP_028687174.1 | 1692 | 76 | |||
| XM_028831342.1 | XP_028687175.1 | 1080 | 76 | |||
| XM_028831343.1 | XP_028687176.1 | 1836 | 76 | |||
| GenBank mRNA | JU322148.1 | - | 1762 | 76 | ||
| JV043462.1 | - | 1733 | 76 | |||
| JV527864.1 | - | 1801 | 76 | |||
Set Membership: |
|