Search
Search

EFTUD2| Gene Symbol: EFTUD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114964.4 | XP_001114964.1 | 1874 | 76 | ||
| XM_015119885.2 | XP_014975371.1 | 2020 | 76 | |||
| XM_015119887.2 | XP_014975373.1 | 2115 | 76 | |||
| XM_015119888.2 | XP_014975374.1 | 1774 | 76 | |||
| XM_028836603.1 | XP_028692436.1 | 2020 | 76 | |||
| GenBank mRNA | JU320559.1 | - | 1749 | 76 | ||
| JU471849.1 | - | 1773 | 76 | |||
| JV665830.1 | - | 1518 | 76 | |||
Set Membership: |
|