Search
Search

CCDC34| Gene Symbol: CCDC34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089845.4 | XP_001089845.2 | 1137 | 117 | ||
| GenBank mRNA | JU496068.1 | - | 480 | 117 | ||
| JV726597.1 | - | 499 | 117 | |||
Set Membership: |
|