Search
Search

DZIP3| Gene Symbol: DZIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015129787.2 | XP_014985273.2 | 1450 | 144 | ||
| XM_015129788.2 | XP_014985274.2 | 1583 | 144 | |||
| XM_015129789.2 | XP_014985275.2 | 2335 | 144 | |||
| XM_015129790.2 | XP_014985276.2 | 1151 | 144 | |||
| XM_015129791.2 | XP_014985277.2 | 1450 | 144 | |||
| XM_015129792.2 | XP_014985278.2 | 1450 | 144 | |||
| XM_015129793.2 | XP_014985279.2 | 1468 | 144 | |||
| XM_028844637.1 | XP_028700470.1 | 1370 | 144 | |||
| XM_028844638.1 | XP_028700471.1 | 1325 | 144 | |||
| XM_028844639.1 | XP_028700472.1 | 718 | 144 | |||
| XM_028844640.1 | XP_028700473.1 | 1220 | 144 | |||
| XM_028844641.1 | XP_028700474.1 | 1228 | 144 | |||
| GenBank mRNA | JU322933.1 | - | 1378 | 144 | ||
Set Membership: |
|